Learn More
Molecular Devices Molecular Devices Microplate Reader, Absorbance, 340-850 nm, 96/384-Well
Catalog No. 5000014464
Encompass_Preferred
The Molecular Devices SpectraMax ABS is an absorbance microplate reader engineered for routine ELISA, microbial growth assays, and nucleic acid and protein quantitation. It provides monochromator-based wavelength selection, precise photometric performance, and temperature control for reliable plate-based absorbance measurements.
  • Monochromator-based wavelength selection, 340-850 nm.
  • Photometric range 0.000-4.000 OD, resolution 0.001 OD.
  • Wavelength selection tunable in 1 nm increments, 2 nm bandwidth.
  • Photometric accuracy < ±0.006 OD ±1.0% (0-3 OD), precision < ±0.003 OD ±1.0% (0-3 OD).
  • Read time 8 seconds for a 96-well plate (kinetic interval).
  • Compatible with 96-well and 384-well plates; supports flat, round, and half-area wells.
  • Temperature control from 5°C above ambient to 45°C, uniformity ±0.75°C at 37°C.
  • Dimensions 8 x 12 x 16.3 in, weight 25 lb; power 100-240 Vac, < 250 W.
Catalog No. 50-000-14464 Supplier Molecular Devices Supplier No. ABS
Add to Cart
Add to Cart
Request a Quote
The Molecular Devices SpectraMax ABS is an absorbance microplate reader engineered for routine ELISA, microbial growth assays, and nucleic acid and protein quantitation. It provides monochromator-based wavelength selection, precise photometric performance, and temperature control for reliable plate-based absorbance measurements.
  • Monochromator-based wavelength selection, 340-850 nm.
  • Photometric range 0.000-4.000 OD, resolution 0.001 OD.
  • Wavelength selection tunable in 1 nm increments, 2 nm bandwidth.
  • Photometric accuracy < ±0.006 OD ±1.0% (0-3 OD), precision < ±0.003 OD ±1.0% (0-3 OD).
  • Read time 8 seconds for a 96-well plate (kinetic interval).
  • Compatible with 96-well and 384-well plates; supports flat, round, and half-area wells.
  • Temperature control from 5°C above ambient to 45°C, uniformity ±0.75°C at 37°C.
  • Dimensions 8 x 12 x 16.3 in, weight 25 lb; power 100-240 Vac, < 250 W.