Learn More
Thomas Scientific Tube Adapter, holds 96 x 0.2ml or 8 and 12 position PCR strips
SDP
Catalog No. 501459069
Encompass
Benchmark's PlateFuge™ is the first "mini" sized centrifuge with a swing out rotor for microplates. Centrifuging plates before PCR ensures all reactants are in the bottom of the wells for proper concentrations and improved yields. In spite of its remarkably small footprint (9x10 in.), the PlateFuge™ includes a uniquely designed, swing out rotor (patent pending) capable of securing 2 microplates. The easily accessible rotor chamber includes two plate carriers that rest at a 75° angle. This allows plates to be inserted confidently without sealing tapes or caps. Upon closing the lid of the centrifuge, the rotor accelerates and the centrifugal force "swings" the plates into a horizontal position. Any droplets on the walls of the plate quickly concentrate into the well bottoms. With a g-force of over 400 xg, most samples can be spun down in less than 20 seconds. To end a run, simply open the lid and the automatic, electric brake brings the rotor to a
Catalog No. 50-145-9069 Supplier Thomas Scientific Supplier No. 1199K64
Benchmark's PlateFuge™ is the first "mini" sized centrifuge with a swing out rotor for microplates. Centrifuging plates before PCR ensures all reactants are in the bottom of the wells for proper concentrations and improved yields. In spite of its remarkably small footprint (9x10 in.), the PlateFuge™ includes a uniquely designed, swing out rotor (patent pending) capable of securing 2 microplates. The easily accessible rotor chamber includes two plate carriers that rest at a 75° angle. This allows plates to be inserted confidently without sealing tapes or caps. Upon closing the lid of the centrifuge, the rotor accelerates and the centrifugal force "swings" the plates into a horizontal position. Any droplets on the walls of the plate quickly concentrate into the well bottoms. With a g-force of over 400 xg, most samples can be spun down in less than 20 seconds. To end a run, simply open the lid and the automatic, electric brake brings the rotor to a