Learn More
Chemglass Life Sciences Adapter, Inert Gas, 29/42 Inner Joint
SDP
Catalog No. CG106402
Encompass_Preferred
Adapter, Inert Gas, 29/42 Inner Joint. Adapter has two hose connections 180º from one another to permit a flow of inert gas over the apparatus. Hose connection has an O.D. of 10mm at the largest serration.
Catalog No. CG106402 Supplier Chemglass Life Sciences Supplier No. CG106402
Adapter, Inert Gas, 29/42 Inner Joint. Adapter has two hose connections 180º from one another to permit a flow of inert gas over the apparatus. Hose connection has an O.D. of 10mm at the largest serration.