Learn More
Chemglass Life Sciences Adapter, Inlet, 19/22 Inner Joint, Bent 90°, 75mm Length Below Joint
SDP
Supplier:  Chemglass Life Sciences CG1014E19
Encompass_Preferred
Adapter, Inlet, 19/22 Inner Joint, Bent 90¿, 75mm Length Below Joint. Allows introduction of reactants below the interior surface. Hose connection has a O.D. of 10mm at the largest serration and is bent downward at a 90¿ angle to the inner joint.
Catalog No. CG1014E19