Learn More
Chemglass Life Sciences Adapter, Inlet, 24/40 Inner Joint, Hose Connection
SDP
Catalog No. CG104601
Encompass_Preferred
Adapter, Inlet, 24/40 Inner Joint, Hose Connection. With a serrated hose connection for vacuum or for the introduction of gases. The #7 Chem-Thread at the top is for a vacuum tight seal of plain stem thermometers or any other tube having an O.D. between 4 and 7mm. Serrated hose connection has an O.D. of 10mm at the largest serration. Supplied complete with a compression cap and Viton o-ring.
Catalog No. CG104601 Supplier Chemglass Life Sciences Supplier No. CG104601
Adapter, Inlet, 24/40 Inner Joint, Hose Connection. With a serrated hose connection for vacuum or for the introduction of gases. The #7 Chem-Thread at the top is for a vacuum tight seal of plain stem thermometers or any other tube having an O.D. between 4 and 7mm. Serrated hose connection has an O.D. of 10mm at the largest serration. Supplied complete with a compression cap and Viton o-ring.