Learn More
Chemglass Life Sciences Adapter, Gas Inlet, 24/40 Joint Size, Hose Connection
SDP
Catalog No. CG106001
Encompass_Preferred
Adapter, Gas Inlet, 24/40 Joint Size, Hose Connection. Gas inlet adapter with a tube extending 200mm below the inner joint. Permits gas to be introduced at the bottom of larger reaction vessels or flasks. With serrated hose connection having an O.D. of 10mm at the largest serration.
Catalog No. CG106001 Supplier Chemglass Life Sciences Supplier No. CG106001
Adapter, Gas Inlet, 24/40 Joint Size, Hose Connection. Gas inlet adapter with a tube extending 200mm below the inner joint. Permits gas to be introduced at the bottom of larger reaction vessels or flasks. With serrated hose connection having an O.D. of 10mm at the largest serration.