Learn More
Chemglass Life Sciences Adapter, Inlet, 29/42 Inner Joint, Bent 90°, 200mm Length Below Joint
SDP
Catalog No. CG1014E02
Encompass_Preferred
Adapter, Inlet, 29/42 Inner Joint, Bent 90¿, 200mm Length Below Joint. Allows introduction of reactants below the interior surface. Hose connection has a O.D. of 10mm at the largest serration and is bent downward at a 90¿ angle to the inner joint.
Catalog No. CG1014E02 Supplier Chemglass Life Sciences Supplier No. CG1014E02
Adapter, Inlet, 29/42 Inner Joint, Bent 90¿, 200mm Length Below Joint. Allows introduction of reactants below the interior surface. Hose connection has a O.D. of 10mm at the largest serration and is bent downward at a 90¿ angle to the inner joint.