Learn More
Chemglass Life Sciences Adapter, Inlet, 14/20 Inner Joint, Bent 90°, 75mm Length Below Joint
SDP
Catalog No. 50872837
Encompass_Preferred
Adapter, Inlet, 14/20 Inner Joint, Bent 90¿, 75mm Length Below Joint. Allows introduction of reactants below the interior surface. Hose connection has a O.D. of 10mm at the largest serration and is bent downward at a 90¿ angle to the inner joint.
Catalog No. 50-872-837 Supplier Chemglass Life Sciences Supplier No. CG1014E14
Adapter, Inlet, 14/20 Inner Joint, Bent 90¿, 75mm Length Below Joint. Allows introduction of reactants below the interior surface. Hose connection has a O.D. of 10mm at the largest serration and is bent downward at a 90¿ angle to the inner joint.