Learn More
Chemglass Life Sciences Adapter, Inlet, 45/50 Inner Joint, 90°
SDP
Catalog No. CG101445
Encompass_Preferred
Adapter, Inlet, 45/50 Inner Joint, Bent 90¿. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration and is bent downward at a 90¿ angle to the inner joint.
Catalog No. CG101445 Supplier Chemglass Life Sciences Supplier No. CG101445
Adapter, Inlet, 45/50 Inner Joint, Bent 90¿. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration and is bent downward at a 90¿ angle to the inner joint.