Learn More
Chemglass Life Sciences Adapter, Inlet, 14/20 Outer Joint, 2mm Glass Stpk, Airfree, Schlenk
SDP
Catalog No. 501530963
Encompass_Preferred
Adapter, Inlet, 14/20 Outer Joint, 2mm Glass Stpk, Airfree, Schlenk. Gas inlet adapter with a 2mm glass stopcock having a constant pressure retention device. Outer standard taper joint. Top arm is 9mm O.D. medium wall tubing for use with CG-3022 septum. Overall height of the 14/20 size is approx. 95mm
Catalog No. 50-153-0963 Supplier Chemglass Life Sciences Supplier No. AF050511
Adapter, Inlet, 14/20 Outer Joint, 2mm Glass Stpk, Airfree, Schlenk. Gas inlet adapter with a 2mm glass stopcock having a constant pressure retention device. Outer standard taper joint. Top arm is 9mm O.D. medium wall tubing for use with CG-3022 septum. Overall height of the 14/20 size is approx. 95mm