Learn More
Chemglass Life Sciences Adapter, Inlet, Bleed Tube, 7mm OD X 265mm OAL
SDP
Catalog No. CG107801
Encompass_Preferred
Adapter, Inlet, Bleed Tube, 7mm OD X 265mm OAL. For the introduction of gases or liquids during reactions. Bleed tube is 7mm OD X 265mm overall length with a tapered delivery end and capillary tip.
Catalog No. CG107801 Supplier Chemglass Life Sciences Supplier No. CG107801
Adapter, Inlet, Bleed Tube, 7mm OD X 265mm OAL. For the introduction of gases or liquids during reactions. Bleed tube is 7mm OD X 265mm overall length with a tapered delivery end and capillary tip.