Learn More
Chemglass Life Sciences Adapter, Inlet, 19/22 Inner Joint, 55mm Height
SDP
Catalog No. CG101219
Encompass_Preferred
Adapter, Inlet, 19/22 Inner Joint, 55mm Height. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration.
Catalog No. CG101219 Supplier Chemglass Life Sciences Supplier No. CG101219
Adapter, Inlet, 19/22 Inner Joint, 55mm Height. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration.