Learn More
Chemglass Life Sciences Adapter, Inlet, Vacuum, Minum-Ware
SDP
Catalog No. 50872926
Encompass_Preferred
Adapter, Inlet, Vacuum, Minum-Ware. Adapter has a 14/10 inner joint and #7 Chem-Thread to permit insertion of 5-7mm O.D. thermometers, bleed tubes etc.; and a side hose connection. Supplied complete with compression cap (CG-352-01) and Viton O-ring (CG-305-07).
Catalog No. 50-872-926 Supplier Chemglass Life Sciences Supplier No. MW3301
Adapter, Inlet, Vacuum, Minum-Ware. Adapter has a 14/10 inner joint and #7 Chem-Thread to permit insertion of 5-7mm O.D. thermometers, bleed tubes etc.; and a side hose connection. Supplied complete with compression cap (CG-352-01) and Viton O-ring (CG-305-07).