Learn More
Wilmad Labglass M-W Inlet Adapter w/Bubbler Tube 24/40
Catalog No. 50803709
Encompass
Apparatus is used for the determination of sulfites in food and related products. Assembly consists of a three neck 1000mL boiling flask with 24/40 joints, a 300mm Allihn condenser and a right angle inlet adapter with a bubbler tube. Remaining components are a right angle hose connection adapter and a special 24/40 outlet adapter with a double bend.
Catalog No. 50-803-709 Supplier Wilmad Labglass Supplier No. LG6675104
Apparatus is used for the determination of sulfites in food and related products. Assembly consists of a three neck 1000mL boiling flask with 24/40 joints, a 300mm Allihn condenser and a right angle inlet adapter with a bubbler tube. Remaining components are a right angle hose connection adapter and a special 24/40 outlet adapter with a double bend.