Learn More
Ideal Vacuum Products LLC Adapter KF-25 to Rubber Hose Nozzle, 9.5mm or 3/8in ID, ISO-KF Flange Size NW-25, Stainless Steel
SDP
Catalog No. NC1715566
Encompass
These adapter rubber hose nozzles are KF-25 to rubber hose and fit a 9.5mm or 3/8in ID rubber hose. The flange size follows ISO-KF standards and is NW-25. They are commonly used to make connections between ISO-KF fittings and rubber vacuum hoses. The ISO-KF flanges are often called quick flange fittings because they are quick to assemble. A circumferential clamp and centering ring form the vacuum seal between the connecting flanges
Catalog No. NC1715566 Supplier Ideal Vacuum Products LLC Supplier No. P108705
These adapter rubber hose nozzles are KF-25 to rubber hose and fit a 9.5mm or 3/8in ID rubber hose. The flange size follows ISO-KF standards and is NW-25. They are commonly used to make connections between ISO-KF fittings and rubber vacuum hoses. The ISO-KF flanges are often called quick flange fittings because they are quick to assemble. A circumferential clamp and centering ring form the vacuum seal between the connecting flanges