Learn More
Perkin Elmer US LLC Headspace Adapter Kit - HP 7890/6890/6850/5890
Catalog No. 509042442
Encompass_Preferred
This kit includes two modified septum nut adapters for packed and capillary operation and the instructions for mechanical installation and interfacing of the two instruments. Note: Installation on HP6890 or HP6850 requires a TurboMatrix headspace sampler to HP6890/6850 GC Ready/Start Cable (M0413591) or an HS40 (XL) to HP6890/HP6850 GC Ready/Start Cable (N6100410).
Catalog No. 50-904-2442 Supplier Perkin Elmer US LLC Supplier No. B0505977
This kit includes two modified septum nut adapters for packed and capillary operation and the instructions for mechanical installation and interfacing of the two instruments. Note: Installation on HP6890 or HP6850 requires a TurboMatrix headspace sampler to HP6890/6850 GC Ready/Start Cable (M0413591) or an HS40 (XL) to HP6890/HP6850 GC Ready/Start Cable (N6100410).