Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO ADAPTER M24 FOR 10MM INSERTS P

Catalog No. 502741541 Shop All METTLER TOLEDO Products
Encompass

Adapter M24 for 10 mm inserts PTFE

Catalog No. 50-274-1541 Supplier METTLER TOLEDO Supplier No. 51162381
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adapter M24 for 10 mm inserts PTFE

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.