Learn More
Mettler Toledo Adapter M24 NS14
Catalog No. 01672598 Shop All METTLER TOLEDO Products
Catalog No. 01-672-598 Supplier Mettler Toledo Supplier No. 30988526
Add to Cart
Add to Cart
Offers NS14/15 adapters (also known as adapter sleeves) for sensors/electrodes with certain shaft diameters
  • Adapter required to connect drying ovens to METTLER TOLEDO EVA Karl Fischer titrators.
  • Adapter is particularly useful in laboratories when you want to use a sensor whose shaft diameter does not match the titration vessel standard port, it avoids having to buy new vessels or inappropriate sensors.
  • It helps maintain proper sealing and alignment, which is essential for accurate titration (or pH / electrochemical measurement), preventing leaks, contamination or measurement error.