Learn More
Chemglass Life Sciences 24/40 Outer Joint Adapter
SDP
Catalog No. 50872899
Encompass_Preferred
24/40 Outer Joint Adapter. Adapter allows for 1/8" OD PTFE tubing to be connected to glassware via a standard taper joint. Useful with vacuum manifolds or any other air sensitive transfer applications. Adapter can also be used with standard luer lock stainless steel needles; See CG-3075. Adapters are supplied with a CG-1055-20 female luer to 1/4" - 28 peek fitting. Ferrules, nuts and tubing must be ordered separately.
Catalog No. 50-872-899 Supplier Chemglass Life Sciences Supplier No. CG105513
24/40 Outer Joint Adapter. Adapter allows for 1/8" OD PTFE tubing to be connected to glassware via a standard taper joint. Useful with vacuum manifolds or any other air sensitive transfer applications. Adapter can also be used with standard luer lock stainless steel needles; See CG-3075. Adapters are supplied with a CG-1055-20 female luer to 1/4" - 28 peek fitting. Ferrules, nuts and tubing must be ordered separately.