Learn More
Chemglass Life Sciences PTFE Microwave Vial Rotary Evap Adapter, 29/42
SDP
Catalog No. 50873521
Encompass_Preferred
PTFE Microwave Vial Rotary Evap Adapter, 29/42. PTFE adapter allows convenient connection of a Microwave vial to a Rotary Evaporator Bump Trap having a standard taper joint size. Lower portion incorporates a threaded black Acetal compression cap that is slotted to accept the flange fitting on the vial.
Catalog No. 50-873-521 Supplier Chemglass Life Sciences Supplier No. CG1318A12
PTFE Microwave Vial Rotary Evap Adapter, 29/42. PTFE adapter allows convenient connection of a Microwave vial to a Rotary Evaporator Bump Trap having a standard taper joint size. Lower portion incorporates a threaded black Acetal compression cap that is slotted to accept the flange fitting on the vial.