Learn More
Chemglass Life Sciences Adapters, Airfree, Schlenk, #15 O-Ring Joint To 3/4" Tubing, Bent 90°
SDP
Catalog No. 50872813
Encompass_Preferred
Adapters, Airfree, Schlenk, #15 O-Ring Joint To 3/4" Tubing, Bent 90. Adapters for use with AF-0060 thru AF-0062 vacuum manifolds that permit the manifold to be utilized with a multitude of standard glassware or tubing products. The AF-0070 adapters are to be used with either a Cajon fitting on the main ports or at the ends of the manifold with heavy wall rubber vacuum tubing. Serrated hose connection adapters list the OD of the hose connection at the middle ring. Each adapter is supplied complete with one #116 Viton o-ring. Adapters are to be attached to the manifold via a CG-150-05 Stainless Steel pinch clamp.
Catalog No. 50-872-813 Supplier Chemglass Life Sciences Supplier No. AF007020
Adapters, Airfree, Schlenk, #15 O-Ring Joint To 3/4" Tubing, Bent 90. Adapters for use with AF-0060 thru AF-0062 vacuum manifolds that permit the manifold to be utilized with a multitude of standard glassware or tubing products. The AF-0070 adapters are to be used with either a Cajon fitting on the main ports or at the ends of the manifold with heavy wall rubber vacuum tubing. Serrated hose connection adapters list the OD of the hose connection at the middle ring. Each adapter is supplied complete with one #116 Viton o-ring. Adapters are to be attached to the manifold via a CG-150-05 Stainless Steel pinch clamp.