Learn More
Ace Glass, Inc. ADAPTER NO.50 ADAPTER NO.50
SDP
Catalog No. 50973327
Encompass_Preferred
#50 Ace-Thred adapter for use with umbrella freeze drying apparatus utilizing Ace-Threds for positive, grease-free connections that are easy to manipulate.
Catalog No. 50-973-327 Supplier Ace Glass, Inc. Supplier No. 669650
#50 Ace-Thred adapter for use with umbrella freeze drying apparatus utilizing Ace-Threds for positive, grease-free connections that are easy to manipulate.