Learn More
Cole-Parmer Edwards C105-04-105 Adapter, NW 16 to 3/4" hose barb, 316L SS
Catalog No. NC0497739 Shop All Cole-Parmer Products
Encompass_Preferred
Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring .
Catalog No. NC0497739 Supplier Cole-Parmer Supplier No. 3140173
Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring .