Learn More
Ace Glass, Inc. Adapter, nylon, filter flask to funnel, #25-#25
SDP
Catalog No. 50971339
Encompass_Preferred
#25 - #25 Nylon adapter connects the filter flask to the funnel on the ACE borosilicate glass filtration apparatus that accomm ODates 25mm membrane filters.
Catalog No. 50-971-339 Supplier Ace Glass, Inc. Supplier No. 370006
#25 - #25 Nylon adapter connects the filter flask to the funnel on the ACE borosilicate glass filtration apparatus that accomm ODates 25mm membrane filters.