Learn More
Chemglass Life Sciences Adapter, Offset, 24/40 Joint Size, 180mm H x 40mm W
SDP
Catalog No. CG103301
Encompass_Preferred
Adapter, Offset, 24/40 Joint Size, 180mm H x 40mm W. With top outer and lower inner standard taper joints. Offset dimension listed is center to center of the joints.
Catalog No. CG103301 Supplier Chemglass Life Sciences Supplier No. CG103301
Adapter, Offset, 24/40 Joint Size, 180mm H x 40mm W. With top outer and lower inner standard taper joints. Offset dimension listed is center to center of the joints.