Learn More
Ace Glass, Inc. Adapter, offset, #15 Ace-Thred top, 14mm OD tube bottom w/O-ring grove.
SDP
Catalog No. 50971815
Encompass_Preferred
Catalog No. 50-971-815 Supplier Ace Glass, Inc. Supplier No. 526912