Learn More
Chemglass Life Sciences Adapter, Outer Luer Joint to 20-400 GPI Thread
SDP
Catalog No. 50872880
Encompass_Preferred
Adapter, Outer Luer Joint to 20-400 GPI Thread. Adapter with outer luer joint provides a convenient way to connect solid phase extraction (SPE) cartridges, tubes or syringes. The adapter provides for easier sample concentration and clean-up while reducing the chances of cross-contamination. Adapters are used for filtration directly into vials. There are three top configurations, with the bottom of all adapters having a 20-400 GPI thread for connection to vials using a CG-1318 Threaded Connector. Design incorporates a drip tube sealed within thread to prevent breakage and are supplied with a 2-1/2” PTFE delivery stem. Hose connection is 10mm OD at the largest serration. ADAPTERS ARE SUPPLIED WITH PTFE DELIVERY STEM.
Catalog No. 50-872-880 Supplier Chemglass Life Sciences Supplier No. CG1049C04
Adapter, Outer Luer Joint to 20-400 GPI Thread. Adapter with outer luer joint provides a convenient way to connect solid phase extraction (SPE) cartridges, tubes or syringes. The adapter provides for easier sample concentration and clean-up while reducing the chances of cross-contamination. Adapters are used for filtration directly into vials. There are three top configurations, with the bottom of all adapters having a 20-400 GPI thread for connection to vials using a CG-1318 Threaded Connector. Design incorporates a drip tube sealed within thread to prevent breakage and are supplied with a 2-1/2” PTFE delivery stem. Hose connection is 10mm OD at the largest serration. ADAPTERS ARE SUPPLIED WITH PTFE DELIVERY STEM.