Learn More
Cole-Parmer Idex Threaded Adapter, PEEK, 0.040" ID, 5/16-24 Flat Bottom (F) to 1/4-28 Flat Bottom (M)
Catalog No. NC2030344 Shop All Cole-Parmer Products
Encompass
Made from PEEK or PCTFE, these adapters are durable and have excellent chemical resistance.
Catalog No. NC2030344 Supplier Cole-Parmer Supplier No. 0202445
Made from PEEK or PCTFE, these adapters are durable and have excellent chemical resistance.