Learn More
Cole-Parmer ADAPTER P-692 1EA
Supplier:  Cole-Parmer 0202369
Encompass
Idex P-692 Adapter Fitting, for P-794, Natural PEEK, 0.020" Bore, 0.020" to 0.030" Hose Barb x Male 1/4-28 Flat Bottom; 1/EA
Catalog No. NC1952874