Learn More
DSI LLC Adapter, PEEK, 10-32 female to 1/4''-28 male
Catalog No. 502254473
Encompass_Preferred
Adapter, PEEK, 10-32 female to 1/4''-28 male
Catalog No. 50-225-4473 Supplier DSI LLC Supplier No. JR0617
Adapter, PEEK, 10-32 female to 1/4''-28 male