Learn More
Chemglass Life Sciences 12mm pH Electrode Adapter, 24/40
SDP
Catalog No. CG1872A01
Encompass_Preferred
Adapter permits CG-1872 series pH electrode(12mm OD) to be inserted either directly into smaller reactors or down through a CG-1872-B baffle.CG-1872-A-01 compression 24/40 joint adapters are to be used with the CG-1872-01 and CG-1872-02 electrodes/probes. The adapter allows use of these probes with reactors up to 4L having lids with a 24/40 neck joint size.Code CG-1872-A-10 is for use CG-1872-03 probes with the open bottom baffles for 10L and larger reactors.
Catalog No. CG1872A01 Supplier Chemglass Life Sciences Supplier No. CG1872A01
Adapter permits CG-1872 series pH electrode(12mm OD) to be inserted either directly into smaller reactors or down through a CG-1872-B baffle.CG-1872-A-01 compression 24/40 joint adapters are to be used with the CG-1872-01 and CG-1872-02 electrodes/probes. The adapter allows use of these probes with reactors up to 4L having lids with a 24/40 neck joint size.Code CG-1872-A-10 is for use CG-1872-03 probes with the open bottom baffles for 10L and larger reactors.