Learn More
AVANTOR MASTERFLEX (VWR LLC) Parker Hannifin Compression threaded adapter, PFA, 1/4"" tubing OD x 1/2"" female NPT
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3132071
Encompass
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Catalog No. 50-631-913