Learn More
Mettler Toledo Adapter Plate KFV GL80
Catalog No. 01672599 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Adapter Plate
Catalog No. Type
01-672-599 Adapter Plate
1 options
Catalog No. 01-672-599 Supplier Mettler Toledo Supplier No. 30988534
Add to Cart
Add to Cart
Allow a titration vessel (or cell) to be mounted / fit properly in the titrator base, used with GL80-series titration vessels / cells
  • Its main purpose is to allow a titration vessel (or cell) to be mounted / fit properly in the titrator base effectively adapting the cell so the system can perform Karl-Fischer titration (water content analysis).
  • The GL80 vessels are part of the product family for coulometric Karl-Fischer titration and are compatible with the EVA titrator series from Mettler Toledo.
  • Chemical or pharmaceutical labs performing coulometric Karl-Fischer titration for moisture/water content analysis.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter Plate