Learn More
Mettler Toledo Adapter Plate Seal KF GL80
Catalog No. 01672600 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Adapter Plate
Catalog No. Type
01-672-600 Adapter Plate
1 options
Catalog No. 01-672-600 Supplier Mettler Toledo Supplier No. 30988535
Add to Cart
Add to Cart
A spare-part/accessory for titration systems, it is the seal component for the adapter plate used in some titration vessels/cells.
  • Adapter plate seal KF GL80. METTLER TOLEDO EVA Coulometric Karl Fischer Titrators.
  • Helps mount or adapt the vessel or electrodes properly on the titrator base.
  • The Adapter plate seal KF thus ensures a proper seal preventing leaks, ingress of moisture from outside, or contamination.
  • Without a good seal, accuracy of KF titration (which is very sensitive to water ingress) would be compromised.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter Plate