Learn More
Chemglass Life Sciences Plug Adapter, 3-Prong, NEMA 5-15R to IEC 60320-C14
SDP
Catalog No. NC1641696
Encompass
Plug Adapter, 3-Prong, NEMA 5-15R to IEC 60320-C14. NEMA 5-15R to IEC 60320-C14 Plug AdapterRated up to 10A 125V. Fully molded rugged construction. For use with Finesse controllers. CAUTION: This plug adapter does NOT transform voltage. Verify your device or power supply is within the output range before use or damage will occur.
Catalog No. NC1641696 Supplier Chemglass Life Sciences Supplier No. CG321111
Plug Adapter, 3-Prong, NEMA 5-15R to IEC 60320-C14. NEMA 5-15R to IEC 60320-C14 Plug AdapterRated up to 10A 125V. Fully molded rugged construction. For use with Finesse controllers. CAUTION: This plug adapter does NOT transform voltage. Verify your device or power supply is within the output range before use or damage will occur.