Learn More
Chemglass Life Sciences Adapter, 80mm Flange to 4" Sanitary, PTFE
SDP
Catalog No. 501310988
Encompass_Preferred
Adapter, 80mm Flange to 4" Sanitary, PTFE. 80mm Adapter to sanitary fitting adapter. Provides a convenient way to connect powder charging bags / bulk addition bags and other sanitary fitting accessories to the addition port on Chemglass reaction systems. Approximate overall length is 2.5¿. Custom sizes and materials are available. NOTE: Clamp and Seals Must Be Purchased Separately.
Catalog No. 50-131-0988 Supplier Chemglass Life Sciences Supplier No. CG1003B10
Adapter, 80mm Flange to 4" Sanitary, PTFE. 80mm Adapter to sanitary fitting adapter. Provides a convenient way to connect powder charging bags / bulk addition bags and other sanitary fitting accessories to the addition port on Chemglass reaction systems. Approximate overall length is 2.5¿. Custom sizes and materials are available. NOTE: Clamp and Seals Must Be Purchased Separately.