Learn More
Ace Glass, Inc. Adapter, PTFE, filter flask to funnel, #25-#25
SDP
Catalog No. 50971338
Encompass
#25 - #25 PTFE adapter connects the filter flask to the funnel on the ACE borosilicate glass filtration apparatus that accomm ODates 25mm membrane filters.
Catalog No. 50-971-338 Supplier Ace Glass, Inc. Supplier No. 370005
#25 - #25 PTFE adapter connects the filter flask to the funnel on the ACE borosilicate glass filtration apparatus that accomm ODates 25mm membrane filters.