Learn More
Ace Glass, Inc. Adapter, filter, PTFE, #80 Ace-Thred to 3/8in female NPT, 100micron polyethylene filter, 350 micron polypropylene filter, retaining ring, FETFE O-ring
SDP
Supplier:  Ace Glass, Inc. 583882
Encompass_Preferred
Catalog No. 50-978-494