Learn More
Med Vet International QUIKVUE PLUS SMARTPHONE EYE IMAGING ADAPTER (2ND GENERATION), J1554M, Each
Catalog No. 5000495059
Encompass_Preferred
It is now easy to obtain high-quality, magnified images of the eye to aid in Ophthalmology diagnosis. The QuikVue Plus is a smartphone adaptor designed especially for anterior segment imaging. It provides a 10x magnification that can be readily saved for the patient. Features: Integrated Yellow Filter • Flippable yellow filter for enhanced fluorescein images with cobalt blue illumination. Useful for cornea ulcer imaging, tear film break-up, time examination, etc. Smartphone Compatibility • Flexible air cushion design allows QuikVue Plus to attach to most smartphones. Battery • 3.7V, 320mAh rechargeable Li-Ion battery w/Type-C USB Cable • 5 hours working time Image Capture • Still image and video QuikVue Plus • Dimensions: 2.36" x 1.57" x 1.57" • Net Weight: 1.41 oz. (40g) • Working Distance: 15~20mm
Catalog No. 50-004-95059 Supplier Med Vet International Supplier No. JOR1554M
It is now easy to obtain high-quality, magnified images of the eye to aid in Ophthalmology diagnosis. The QuikVue Plus is a smartphone adaptor designed especially for anterior segment imaging. It provides a 10x magnification that can be readily saved for the patient. Features: Integrated Yellow Filter • Flippable yellow filter for enhanced fluorescein images with cobalt blue illumination. Useful for cornea ulcer imaging, tear film break-up, time examination, etc. Smartphone Compatibility • Flexible air cushion design allows QuikVue Plus to attach to most smartphones. Battery • 3.7V, 320mAh rechargeable Li-Ion battery w/Type-C USB Cable • 5 hours working time Image Capture • Still image and video QuikVue Plus • Dimensions: 2.36" x 1.57" x 1.57" • Net Weight: 1.41 oz. (40g) • Working Distance: 15~20mm