Learn More
Thomas Scientific Reducing Adapters for use in 15mL tubes, Sterile
SDP
Catalog No. 501457902
Encompass_Preferred
The SureStrain™ Premium Cell Strainer system is a fast and easy-to-use solution for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh that is available in three color-coded porosities: 40μm (blue), 70μm (white), and 100μm (yellow). A tiered filtration process is made possible by the SureStrain’s unique interlocking stackable design. SureStrain’s molded grip and unique geometry ensure aseptic handling and eliminate the contamination risks associated with fingers contacting the filtered suspension, mesh and/or interior surfaces. The sterile strainer can even be removed from the special blister packaging without touching its interior surface or mesh. The advanced frame/housing design also contributes to enhanced performance and reduced processing time, by eliminating the "air-lock" effect. Improved aeration allows air to escape while filtrate is entering the tube and prevents messy spillover
Catalog No. 50-145-7902 Supplier Thomas Scientific Supplier No. 1181X55
The SureStrain™ Premium Cell Strainer system is a fast and easy-to-use solution for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh that is available in three color-coded porosities: 40μm (blue), 70μm (white), and 100μm (yellow). A tiered filtration process is made possible by the SureStrain’s unique interlocking stackable design. SureStrain’s molded grip and unique geometry ensure aseptic handling and eliminate the contamination risks associated with fingers contacting the filtered suspension, mesh and/or interior surfaces. The sterile strainer can even be removed from the special blister packaging without touching its interior surface or mesh. The advanced frame/housing design also contributes to enhanced performance and reduced processing time, by eliminating the "air-lock" effect. Improved aeration allows air to escape while filtrate is entering the tube and prevents messy spillover