Learn More
Kuhner Shaker Inc ADAPTER RING TO MOUNT THE..TOM
Catalog No. 502570082
Encompass
Adapter ring to mount the..TOM cell culture adapter lid onto..a 50 mL Tube....needs cell culture adapter lid..125 mL, 250 mL, 500 mL and 1000 mL..Corning cell culture flasks;..not autoclavable
Catalog No. 50-257-0082 Supplier Kuhner Shaker Inc Supplier No. SMTO00007
Adapter ring to mount the..TOM cell culture adapter lid onto..a 50 mL Tube....needs cell culture adapter lid..125 mL, 250 mL, 500 mL and 1000 mL..Corning cell culture flasks;..not autoclavable