Learn More
Chemglass Life Sciences Shower Head Adapter
SDP
Catalog No. 501531253
Encompass_Preferred
Shower Head Adapter. Adapter attaches to the top of the exhaust gas outlet column and has an all glass shower head to disperse the scrubbing fluid down into the absorption column. Inlet connection is 1/2 Swagelok tubing for use with CG-1975-20 compression fitting.
Catalog No. 50-153-1253 Supplier Chemglass Life Sciences Supplier No. CG183016
Shower Head Adapter. Adapter attaches to the top of the exhaust gas outlet column and has an all glass shower head to disperse the scrubbing fluid down into the absorption column. Inlet connection is 1/2 Swagelok tubing for use with CG-1975-20 compression fitting.