Learn More
Beckman Coulter Adapter Sleeve, Polypropylene, Bottle, 62 mm dia (qty. 4)
Catalog No. NC1157907
Encompass
Pale Red; Tube Volumes: 230 mL Conical; Max Tube Diameter: 62 mm; Number of Places in Adapter: 1; Number of Tubes in Rotor: 4; Adapter for Tubes in Beckman Coulter Rotors. For use in High Performance and High Capacity Rotors:
Catalog No. NC1157907 Supplier Beckman Coulter Supplier No. 356985
Pale Red; Tube Volumes: 230 mL Conical; Max Tube Diameter: 62 mm; Number of Places in Adapter: 1; Number of Tubes in Rotor: 4; Adapter for Tubes in Beckman Coulter Rotors. For use in High Performance and High Capacity Rotors: