Learn More
Chemglass Life Sciences Adapter, Solid Addition, Merlic, 14/20 Joint Size, Airfree, Schlenk
SDP
Catalog No. AF050301
Encompass_Preferred
Adapter, Solid Addition, Merlic, 14/20 Joint Size, Airfree, Schlenk. Adapter permits addition of solids into multi-neck flasks in a dry box, or addition of non-air sensitive compounds into flasks while purging with an inert atmosphere. Can handle from 50mg to 1 gram of solids and is made with a drip tube to avoid getting the solids on the joint or the wall of the flask.** Design Suggested By Dr. Craig Merlic, University Of California, Los Angeles, CA
Catalog No. AF050301 Supplier Chemglass Life Sciences Supplier No. AF050301
Adapter, Solid Addition, Merlic, 14/20 Joint Size, Airfree, Schlenk. Adapter permits addition of solids into multi-neck flasks in a dry box, or addition of non-air sensitive compounds into flasks while purging with an inert atmosphere. Can handle from 50mg to 1 gram of solids and is made with a drip tube to avoid getting the solids on the joint or the wall of the flask.** Design Suggested By Dr. Craig Merlic, University Of California, Los Angeles, CA