Learn More
Ace Glass, Inc. Adapter, splash, 14/20 top, 14/20 bottom, porosity A
SDP
Catalog No. 50971788
Encompass_Preferred
Firestone splash guard adapter with medium length joints that will accept full length joints. Internal porosity A (147-174 micron) fritted disc that assures no carry-over in the event of pot splash-up. Overall length is kept to a minimum to positively effect distillation.
Catalog No. 50-971-788 Supplier Ace Glass, Inc. Supplier No. 525743
Firestone splash guard adapter with medium length joints that will accept full length joints. Internal porosity A (147-174 micron) fritted disc that assures no carry-over in the event of pot splash-up. Overall length is kept to a minimum to positively effect distillation.