Learn More
Chemglass Life Sciences Adapter, Splash Guard, Top: 14/20, Lower: 14/20
SDP
Catalog No. 50872851
Encompass_Preferred
Adapter, Splash Guard, Top: 14/20, Lower: 14/20. Adapter with a choice of full or medium length joints & when placed between the distilling flask & column prevents splash-ups. Overall height is minimal to aid in effective distillation.
Catalog No. 50-872-851 Supplier Chemglass Life Sciences Supplier No. CG104101
Adapter, Splash Guard, Top: 14/20, Lower: 14/20. Adapter with a choice of full or medium length joints & when placed between the distilling flask & column prevents splash-ups. Overall height is minimal to aid in effective distillation.