Learn More
Chemglass Life Sciences Adapter, Splash Guard, Top: 24/40, Lower: 24/40
SDP
Catalog No. 50872853
Encompass_Preferred
Adapter, Splash Guard, Top: 24/40, Lower: 24/40. Adapter with a choice of full or medium length joints & when placed between the distilling flask & column prevents splash-ups. Overall height is minimal to aid in effective distillation.
Catalog No. 50-872-853 Supplier Chemglass Life Sciences Supplier No. CG104103
Adapter, Splash Guard, Top: 24/40, Lower: 24/40. Adapter with a choice of full or medium length joints & when placed between the distilling flask & column prevents splash-ups. Overall height is minimal to aid in effective distillation.