Learn More
Chemglass Life Sciences Adapter, Splash Guard, Top: 29/42, Lower: 29/42
SDP
Catalog No. 50872855
Encompass_Preferred
Adapter, Splash Guard, Top: 29/42, Lower: 29/42. Adapter with a choice of full or medium length joints & when placed between the distilling flask & column prevents splash-ups. Overall height is minimal to aid in effective distillation.
Catalog No. 50-872-855 Supplier Chemglass Life Sciences Supplier No. CG104105
Adapter, Splash Guard, Top: 29/42, Lower: 29/42. Adapter with a choice of full or medium length joints & when placed between the distilling flask & column prevents splash-ups. Overall height is minimal to aid in effective distillation.