Learn More
Grainger Adapter SS 1 1/2 in Socket 29 PSI
Catalog No. 502753043
Encompass_Preferred
The Grainger 14P010 is a 14-inch diameter, 10-inch long, cast iron pulley with a 1-1/8 inch bore and keyway. It has an A-section v-belt groove and is rated for speeds up to 6580 feet per minute. This heavy-duty pulley is made in the USA out of gray cast iron with an oil finish. It can be used with v-belts sized from A24 to A48. The pulley weighs approximately 13 pounds.
Catalog No. 50-275-3043 Supplier Grainger Supplier No. 14P010
The Grainger 14P010 is a 14-inch diameter, 10-inch long, cast iron pulley with a 1-1/8 inch bore and keyway. It has an A-section v-belt groove and is rated for speeds up to 6580 feet per minute. This heavy-duty pulley is made in the USA out of gray cast iron with an oil finish. It can be used with v-belts sized from A24 to A48. The pulley weighs approximately 13 pounds.