Learn More
Ace Glass, Inc. Adapter, SS, 1/2in beaded pipe to 1/2in male NPT
SDP
Catalog No. 50976621
Encompass
304 Stainless Steel adapter connects 1/2 beaded pipe to 1/2 male NPT fittings, gauges, and flanges.
Catalog No. 50-976-621 Supplier Ace Glass, Inc. Supplier No. 887002
304 Stainless Steel adapter connects 1/2 beaded pipe to 1/2 male NPT fittings, gauges, and flanges.